AMBH2D6F0419 Ambeed, Inc.

Ethyl 4,5,6,7-tetrahydrothieno[3,2-c]pyridine-2-carboxylate

  • MDL number MFCD10566074
  • Cas Number : 1080026-94-3
  • Molecular Weight : 211.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1080026-94-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1080026-94-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart