AMBH2D6F041D Ambeed, Inc.

Ethyl 1-methyl-4-nitro-1H-imidazole-2-carboxylate

  • MDL number MFCD03789110
  • Cas Number : 109012-23-9
  • Molecular Weight : 199.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/109012-23-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/109012-23-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart