AMBH2D6F0434 Ambeed, Inc.

BML-284

  • MDL number MFCD09908087
  • Cas Number : 853220-52-7
  • Molecular Weight : 350.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/853220-52-7.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/853220-52-7.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart