AMBH2D6F0475 Ambeed, Inc.

5-Bromo-N-methylpyrimidin-4-amine

  • MDL number MFCD03861147
  • Cas Number : 56181-38-5
  • Molecular Weight : 188.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/56181-38-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/56181-38-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart