AMBH2D6F0B39 Ambeed, Inc.

Methyl 1H-pyrrolo[3,2-b]pyridine-3-carboxylate

  • MDL number MFCD09864655
  • Cas Number : 952800-39-4
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/952800-39-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/952800-39-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart