AMBH2D6F0B58 Ambeed, Inc.

Ethyl 4-nitro-1H-pyrazole-3-carboxylate

  • MDL number MFCD00195040
  • Cas Number : 55864-87-4
  • Molecular Weight : 185.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/55864-87-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/55864-87-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart