AMBH2D6F0BA6 Ambeed, Inc.

Imidazo[2,1-b]thiazole-5-carboxylic acid

  • MDL number MFCD09743157
  • Cas Number : 17782-81-9
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/17782-81-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/17782-81-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart