AMBH2D6F0BB2 Ambeed, Inc.

BSI-201

  • MDL number MFCD11110639
  • Cas Number : 160003-66-7
  • Molecular Weight : 292.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/160003-66-7.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/160003-66-7.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart