AMBH2D6F0D08 Ambeed, Inc.

1-Boc-4-Cbz-aminoazepane

  • MDL number MFCD18207774
  • Cas Number : 878630-96-7
  • Molecular Weight : 348.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/878630-96-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/878630-96-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart