AMBH2D6F0D2C Ambeed, Inc.

Xylazole hydrochloride

  • MDL number N/A
  • Cas Number : 123941-49-1
  • Molecular Weight : 240.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/123941-49-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/123941-49-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart