AMBH2D6F0D79 Ambeed, Inc.

6-Amino-8-bromo-1,7-naphthyridine

  • MDL number MFCD09702034
  • Cas Number : 5912-35-6
  • Molecular Weight : 224.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/5912-35-6.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/5912-35-6.html
pictograms GHS07
hstatements H302-H312-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart