AMBH2D6F0DA7 Ambeed, Inc.

Ethyl 5-chloro-1-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD10699191
  • Cas Number : 1172229-72-9
  • Molecular Weight : 188.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1172229-72-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1172229-72-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart