AMBH2D6F0E21 Ambeed, Inc.

1-Boc-3-Ethynylpiperidine

  • MDL number MFCD10698159
  • Cas Number : 664362-16-7
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form White to Yellow Solid or liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 269.6�t 760 mmHg
Product Link https://www.ambeed.com/products/664362-16-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/664362-16-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart