AMBH2D6F0E24 Ambeed, Inc.

2-Ethyl-3-methylbutan-1-ol

  • MDL number MFCD00089635
  • Cas Number : 32444-34-1
  • Molecular Weight : 116.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/32444-34-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/32444-34-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart