AMBH2D6F0E93 Ambeed, Inc.

2-Hydroxypyrimidine-5-carbaldehyde

  • MDL number MFCD10696897
  • Cas Number : 166757-62-6
  • Molecular Weight : 124.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/166757-62-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/166757-62-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart