AMBH2D6F0EB2 Ambeed, Inc.

4-Benzyl-2-phenyloxazol-5(4H)-one

  • MDL number MFCD00157349
  • Cas Number : 5874-61-3
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/5874-61-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/5874-61-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart