AMBH2D6F0EB3 Ambeed, Inc.

(2-Bromomethyl)-4-chloro-1-nitrobenzene

  • MDL number MFCD11848697
  • Cas Number : 31577-25-0
  • Molecular Weight : 250.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/31577-25-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/31577-25-0.html
pictograms GHS07
hstatements H302-H312-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart