AMBH2D6F0F19 Ambeed, Inc.

6-Fluoro-1-methyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid

  • MDL number MFCD00882994
  • Cas Number : 112984-60-8
  • Molecular Weight : 349.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/112984-60-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/112984-60-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart