AMBH2D6F0FDF Ambeed, Inc.

Methyl 1-Boc-3-methyl-4-oxo-piperidine-3-carboxylate

  • MDL number MFCD11506200
  • Cas Number : 193274-53-2
  • Molecular Weight : 271.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/193274-53-2.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/193274-53-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart