AMBH2D6F1011 Ambeed, Inc.

3-Bromo-5-nitro-1-trityl-1H-indazole

  • MDL number MFCD17169854
  • Cas Number : 942189-39-1
  • Molecular Weight : 484.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/942189-39-1.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/942189-39-1.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart