AMBH2D6F102E Ambeed, Inc.

Ethyl 2-(4-(tert-butoxycarbonyl)piperazin-1-yl)thiazole-4-carboxylate

  • MDL number MFCD08445626
  • Cas Number : 867065-53-0
  • Molecular Weight : 341.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/867065-53-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/867065-53-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart