AMBH2D6F1830 Ambeed, Inc.

2-(4-Fluorophenyl)-2,3-dihydro-4(1H)-quinolinone

  • MDL number MFCD09999181
  • Cas Number : 155370-03-9
  • Molecular Weight : 241.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/155370-03-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/155370-03-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart