AMBH2D6F18B2 Ambeed, Inc.

(R)-1,4-Diazabicyclo[4.3.0]nonane

  • MDL number MFCD09752820
  • Cas Number : 96193-27-0
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8°C
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 199.6°C at 760 mmHg
Product Link https://www.ambeed.com/products/96193-27-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/96193-27-0.html
pictograms GHS07
hstatements H227-H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart