AMBH2D6F18DB Ambeed, Inc.

2-(4-Methyl-2-phenyl-1-piperazinyl)-3-pyridinemethanol

  • MDL number MFCD08063948
  • Cas Number : 61337-89-1
  • Molecular Weight : 283.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/61337-89-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/61337-89-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart