AMBH2D6F18F1 Ambeed, Inc.

Dimethyl 2-methylisophthalate

  • MDL number MFCD00229510
  • Cas Number : 28269-31-0
  • Molecular Weight : 208.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/28269-31-0.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/28269-31-0.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart