AMBH2D6F1973 Ambeed, Inc.

2-Chloro-6-methylpyridin-4-amine

  • MDL number MFCD00235132
  • Cas Number : 79055-63-3
  • Molecular Weight : 142.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/79055-63-3.html

Safety Information

signalword Danger
pcodes P261-P280-P305+P351+P338-P311
MSDS https://www.ambeed.com/products/79055-63-3.html
pictograms GHS06
hstatements H302+H312-H315-H319-H331-H335
Availability details for
Quantity
  • Availability
  • Add to Cart