AMBH2D6F19E1 Ambeed, Inc.

9,10-Diethoxyanthracene

  • MDL number MFCD00209643
  • Cas Number : 68818-86-0
  • Molecular Weight : 266.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/68818-86-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/68818-86-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart