AMBH2D6F19EE Ambeed, Inc.

2,3-Dibromoisonicotinic acid

  • MDL number MFCD09997708
  • Cas Number : 1020056-98-7
  • Molecular Weight : 280.9000

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1020056-98-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1020056-98-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart