AMBH2D6F1A03 Ambeed, Inc.

(4-Methyl-3-(trifluoromethyl)phenyl)boronic acid

  • MDL number MFCD11042431
  • Cas Number : 947533-94-0
  • Molecular Weight : 203.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/947533-94-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/947533-94-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart