AMBH2D6F1A8A Ambeed, Inc.

4-Amino-N-(2-diethylaminoethyl)benzamide

  • MDL number MFCD00066880
  • Cas Number : 51-06-9
  • Molecular Weight : 235.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/51-06-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/51-06-9.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart