AMBH2D6F1A8C Ambeed, Inc.

tert-Butyl 7-(trifluoromethyl)-3,4-dihydroisoquinoline-2(1H)-carboxylate

  • MDL number MFCD12923110
  • Cas Number : 1257855-77-8
  • Molecular Weight : 301.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8°C
Physical Form Solid or liquid
Shipping Temperature Normal
Country of Origin China
Boiling point 334.8°42.0°C at 760 mmHg
Product Link https://www.ambeed.com/products/1257855-77-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1257855-77-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart