AMBH2D6F1AC5 Ambeed, Inc.

2-Bromo-4-nitroimidazole

  • MDL number MFCD09038915
  • Cas Number : 65902-59-2
  • Molecular Weight : 191.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-182�t N/A mmHg
Purity 97
Flash Point 197.2
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 402.5�t 760 mmHg
Product Link https://www.ambeed.com/products/65902-59-2.html

Safety Information

signalword Warning
pcodes P264-P270-P280-P301+P312+P330-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313-P501
MSDS https://www.ambeed.com/products/65902-59-2.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart