AMBH2D6F1AC8 Ambeed, Inc.

2-Chloro-5-nitroimidazole

  • MDL number MFCD03419295
  • Cas Number : 57531-37-0
  • Molecular Weight : 147.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/57531-37-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/57531-37-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart