AMBH2D6F1ADE Ambeed, Inc.

4-Chloropyridine hydrochloride

  • MDL number MFCD00012829
  • Cas Number : 7379-35-3
  • Molecular Weight : 150.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/7379-35-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/7379-35-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart