AMBH2D6F1AEB Ambeed, Inc.

2-Methacrylamidoacetic acid

  • MDL number MFCD00466786
  • Cas Number : 23578-45-2
  • Molecular Weight : 143.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Flash Point 190.4
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 391.2�t 760 mmHg
Product Link https://www.ambeed.com/products/23578-45-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/23578-45-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart