AMBH2D6F1C42 Ambeed, Inc.

Methyl 6-(3-(adamantan-1-yl)-4-methoxyphenyl)-2-naphthoate

  • MDL number MFCD08063923
  • Cas Number : 106685-41-0
  • Molecular Weight : 426.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/106685-41-0.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/106685-41-0.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart