AMBH2D6F25C1 Ambeed, Inc.

5-(4'-Methyl-[1,1'-biphenyl]-2-yl)-1-trityl-1H-tetrazole

  • MDL number MFCD00665220
  • Cas Number : 124750-53-4
  • Molecular Weight : 478.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 97
Flash Point 366
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 681.6�t 760 mmHg
Product Link https://www.ambeed.com/products/124750-53-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/124750-53-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart