AMBH2D6F28B9 Ambeed, Inc.

Candesartan

  • MDL number MFCD00864463
  • Cas Number : 139481-59-7
  • Molecular Weight : 440.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/139481-59-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/139481-59-7.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart