AMBH2D6F28CF Ambeed, Inc.

Ethyl 4,6-dihydroxy-5-nitronicotinate

  • MDL number MFCD01765414
  • Cas Number : 6317-97-1
  • Molecular Weight : 228.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/6317-97-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/6317-97-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart