AMBH2D6F2910 Ambeed, Inc.

Bis(triphenylphosphine)nickel(II)chloride

  • MDL number MFCD00009592
  • Cas Number : 14264-16-5
  • Molecular Weight : 654.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/14264-16-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/14264-16-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart