AMBH2D6F2916 Ambeed, Inc.

Tris(2,2'-bipyridine)ruthenium dichloride

  • MDL number MFCD00013224
  • Cas Number : 14323-06-9
  • Molecular Weight : 640.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/14323-06-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/14323-06-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart