AMBH2D6F2998 Ambeed, Inc.

3-(2-Bromopropanoyl)spiro[benzo[e][1,3]oxazine-2,1'-cyclohexan]-4(3H)-one

  • MDL number MFCD08458252
  • Cas Number : 158299-05-9
  • Molecular Weight : 352.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/158299-05-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/158299-05-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart