AMBH2D6F29EE Ambeed, Inc.

Ethyl 2-hydrazinyl-2-oxoacetate

  • MDL number MFCD05665958
  • Cas Number : 35196-48-6
  • Molecular Weight : 132.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-52°C
Synonym N/A
Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/35196-48-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/35196-48-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart