AMBH2D6F2A44 Ambeed, Inc.

Tetraethyl ([1,1'-biphenyl]-4,4'-diylbis(methylene))bis(phosphonate)

  • MDL number MFCD01321144
  • Cas Number : 17919-34-5
  • Molecular Weight : 454.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/17919-34-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/17919-34-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart