AMBH2D6F2A5B Ambeed, Inc.

3-Hydroxypyridine-4-carboxaldehyde

  • MDL number MFCD01318565
  • Cas Number : 1849-54-3
  • Molecular Weight : 123.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1849-54-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1849-54-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart