AMBH2D6F2AF5 Ambeed, Inc.

6-Bromo-4,4-dimethyl-1,4-dihydrobenzo[d][1,3]oxazin-2-one

  • MDL number MFCD00866701
  • Cas Number : 21440-97-1
  • Molecular Weight : 256.1000

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/21440-97-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/21440-97-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart