AMBH2D6F2B2C Ambeed, Inc.

Benzo[d]isothiazol-3-amine

  • MDL number MFCD00726295
  • Cas Number : 23031-78-9
  • Molecular Weight : 150.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/23031-78-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/23031-78-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart