AMBH2D6F2B32 Ambeed, Inc.

N,N'-(Hexane-1,6-diyl)bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanamide)

  • MDL number MFCD00134697
  • Cas Number : 23128-74-7
  • Molecular Weight : 636.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/23128-74-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/23128-74-7.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart