AMBH2D6F2B3E Ambeed, Inc.

Ethyl 4-ethoxybenzoate

  • MDL number MFCD00009116
  • Cas Number : 23676-09-7
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 9
Purity 98
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 275�t 760 mmHg
Product Link https://www.ambeed.com/products/23676-09-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/23676-09-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart