AMBH2D6F2B5A Ambeed, Inc.

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-2-one

  • MDL number MFCD11504971
  • Cas Number : 1150271-44-5
  • Molecular Weight : 259.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1150271-44-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1150271-44-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart